Skip to Content
Merck
All Photos(1)

Key Documents

MX1123

Sigma-Aldrich

1-Methylimidazole

Synonym(s):

1-Methylimidazole, N-Methylimidazole, dna

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3-NCH=NCH=CH-
CAS Number:
MDL number:
UNSPSC Code:
12352100
EC Index Number:
210-484-7

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

form

liquid

pH

9.5-10.5 (20 °C, 50 g/L in H2O)

bp

198 °C/1013 hPa

mp

-2 °C

transition temp

flash point 92 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3

InChI key

MCTWTZJPVLRJOU-UHFFFAOYSA-N

Application

1-Methylimidazole can be used as an organocatalyst to synthesize:
  • 4-amino-3,5-dicyano-6-arylphthalates via one-pot, a three-component reaction between arylidenemalononitriles, dialkyl ­acetylenedicarboxylates, and malononitrile.
  • Aromatic and aliphatic carbamates from hydroxamic acids via the Lossen rearrangement.
  • Tetrahydrobenzo[b]pyrans via three-component cyclocondensation reaction between aromatic aldehydes, malononitrile, and dimedone.

It can also be used as:
  • A co-catalyst in copper-catalyzed, greener oxidation of alcohols to carbonyl compounds under aerobic conditions.
  • A solvent/complexing agent in the preparation of CuInS2 n-type semiconductive particles.
  • A co-additive in the synthesis of acid-sensitive epoxides by methyltrioxorhenium-catalyzed epoxidation of alkenes with H2O2.

Analysis Note

Assay (GC)99.0% minDensity at 20°C1.03-1.04g/mLFormClear, liquidWater (H2O)150ppm max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of n-type CuInS2 particles using N-methylimidazole, characterization and growth mechanism
Courtel FM, et al.
Chemistry of Materials, 22(12), 3752-3761 (2010)
1-Methylimidazole-catalyzed regioselective synthesis of highly substituted benzenes
Adib Mehdi, et al.
Synlett, 2007(16), 2497-2500 (2007)
A green synthesis of tetrahydrobenzo [b] pyran derivatives through three-component condensation using N-methylimidazole as organocatalyst
Lian Xi-Zhong, et al.
Monatshefte fur Chemie / Chemical Monthly, 139(2), 129-131 (2008)
Shigekazu Yamazaki
Organic & biomolecular chemistry, 8(10), 2377-2385 (2010-05-08)
An effective method for suppression of ring opening and rearrangement of acid-sensitive epoxides during methyltrioxorhenium(MTO)-catalyzed epoxidation of alkenes with H(2)O(2) by using 1-methylimidazole as a co-additive has been found. The combined use of 3-methylpyrazole and 1-methylimidazole as the additives has
Janelle E Steves et al.
Journal of the American Chemical Society, 135(42), 15742-15745 (2013-10-17)
Cu/TEMPO catalyst systems promote efficient aerobic oxidation of sterically unhindered primary alcohols and electronically activated substrates, but they show reduced reactivity with aliphatic and secondary alcohols. Here, we report a catalyst system, consisting of ((MeO)bpy)Cu(I)(OTf) and ABNO ((MeO)bpy = 4,4'-dimethoxy-2,2'-bipyridine;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service