Skip to Content
Merck
All Photos(2)

Documents

MABT884

Sigma-Aldrich

Anti-TACE Antibody, clone D1(A12)

clone D1(A12), from human(Recombinant)

Synonym(s):

Disintegrin and metalloproteinase domain-containing protein 17, ADAM 17, CD156b, Snake venom-like protease, TNF-alpha convertase, TNF-alpha-converting enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

human (Recombinant)

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D1(A12), monoclonal

species reactivity

human

should not react with

mouse (Kwok, H.F., et al. (2014). Protein Eng. Des. Sel. 27(6):179-190.)

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ADAM17(6868)

General description

Disintegrin and metalloproteinase domain-containing protein 17 (UniProt: P78536; also known as EC:3.4.24.86, ADAM 17, Snake venom-like protease, TNF-alpha convertase, TNF-alpha-converting enzyme, TACE, CD156b) is encoded by the ADAM17 (also known as CSVP, TACE) gene (Gene ID: 6868) in human. TACE is a membrane-bound metalloprotease responsible for solubilizing many pathologically significant membrane substrates. Mature ADAM-family ectodomains contain a globular metalloprotease catalytic domain, a disulfide-dependent disintegrin cysteine rich (Dis-Cys) domain, and, in some cases, an epidermal growth factor (EGF)-like domain. ADAM17 activity is regulated by multiple post-translational modifications. It has three phosphorylation sites on its C-terminal cytoplasmic domain (T735, S791, and (S819). ERK1/2-mediated T735 phosphorylation is reported to regulate ADAM17 maturation and surface expression. TACE displays a narrow endopeptidase specificity. It cleaves the membrane-bound precursor of TNF-alpha to its mature soluble form and is also responsible for the proteolytic release of several other cell-surface proteins, including p75 TNF-receptor, interleukin 1 receptor type II, p55 TNF-receptor, transforming growth factor-alpha, L-selectin, growth hormone receptor, MUC1 and the amyloid precursor protein. Clone D1(A12) exhibits high affinity (Kd = 0.46 nM) for the TACE ectodomain and is the most selectively potent cell-surface ADAM inhibitor. It can independently bind both the complete TACE ectodomain and the isolated catalytic domain (Kd = 5.21 nM). It is shown to be about 5-fold more potent than N-TIMP3 in inhibiting cell surface TACE. Mutations in ADAM17 gene are known to cause inflammatory skin and bowel disease, neonatal, 1 (NISBD1) that is characterized by inflammatory features with neonatal onset, involving the skin, hair, and gut.

Specificity

Cat. No. MABT884 is a recombinant human IgG1 that contains a heavy chain variable (VH) region with affinity towards human TACE noncatalytic Dis-Cys domain and a light chain variable (VL) region that binds human TACE catalytic domain (Tape, C.J., et al. (2011). Proc. Natl. Acad. Sci. U. S. A. 108(14):5578-5583). The VH region of D1(A12) is derived from the scFv named D1 isolated from a phage library generated using pooled lymphocytes from 43 donors (Schofield, D.J., et al. (2007). Genome Biol. 8(11):R254). The D1 VH cDNA was subsequently cloned into a naïve human VL phage-display library for identification and isolation of the hybrid D1(A12) scFv with affinity toward both Dis-Cys domain and the catalytic domain. The D1 VH and A12 VL cDNAs were then cloned into a pBudCE4.1 human IgG1 expression vector for generating the recombinant antibody in HEK293 cells (Tape, C.J., et al. (2011). Proc. Natl. Acad. Sci. USA. 108(14):5578-5583).

Immunogen

Epitope: extracellular domain
See Specificity section.

Application

Detect TACE/ADAM17 using this human monoclonal Anti-TACE, clone D1(A12), Cat. No. MABT884, validated for use in Flow Cytometry and Activity/Function Inhibition.
Inhibits Activity/Function: 6 µg/mL from a representative lot inhibited TACE-mediated CD16 shedding from the surface of stimulated primary human NK cells (Courtesy of Zachary Davis, Ph.D., University of Minnesota, USA).

Inhibits Activity/Function: Representative lots inhibited TACE-mediated substrates shedding from the surface of human cells both in cultures and in mice in vivo (Issuree, P.D., et al. (2013). J. Clin. Invest. 123(2):928-932; Richards, F.M., et al. (2012). PLoS One. 7(7):e40597; Tape, C.J., et al. (2011). Proc. Natl. Acad. Sci. U. S. A. 108(14):5578-5583).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Flow Cytometry in HeLa cells.

Flow Cytometry Analysis: 0.2 µg (2 µg/mL) of this antibody detected TACE expression on the surface of one million HeLa cells.

Target description

93.02/91.12/68.22 kDa (prepro-/pro-/mature form) calculated.

Physical form

Format: Purified
Purified recombinant human IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathalie Burg et al.
JCI insight, 9(11) (2024-06-10)
In rheumatoid arthritis, inflammatory mediators extravasate from blood into joints via gaps between endothelial cells (ECs), but the contribution of ECs is not known. Sphingosine 1-phosphate receptor 1 (S1PR1), widely expressed on ECs, maintains the vascular barrier. Here, we assessed
Anders W Matson et al.
Journal for immunotherapy of cancer, 12(7) (2024-07-26)
Natural killer (NK) cells are being extensively studied as a cell therapy for cancer. These cells are activated by recognition of ligands and antigens on tumor cells. Cytokine therapies, such as IL-15, are also broadly used to stimulate endogenous and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service