Skip to Content
Merck
All Photos(1)

Key Documents

MAB8254

Sigma-Aldrich

Anti-Influenza A Antibody, H-3 Antigen, clone 30-2F11-F7-A5

clone 30-2F11-F7-A5, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

30-2F11-F7-A5, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Recognizes Influenza A H3 antigen. Reacts strongly with the following H3N2 strains: A/Wuhan/396/95; A/Johannesburg/33/94;

A/Shandung/93;

A/Shanghai/16/89; All CA Strains 1986 to Present.

No reactivity shown with the following H1N1 strains: A/Taiwan/1/86;A/Texas/36/91; A/USSR/90/77; All CA strains 1986 to present.

No reactivity shown to Influenza B strains.

Immunogen

Epitope: H-3 Antigen
Influenza blend.

Application

Detect Influenza A using this Anti-Influenza A Antibody, H-3 Antigen, clone 30-2F11-F7-A5 validated for use in IF.
Indirect Immunofluorescence
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Influenza Control Slides, Catalogue Number 5010-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canine H3N8 influenza virus infection in dogs and mice.
W L Castleman,J R Powe,P C Crawford,E P J Gibbs,E J Dubovi,R O Donis,D Hanshaw
Veterinary Pathology null
Ning Chai et al.
PLoS pathogens, 12(6), e1005702-e1005702 (2016-06-29)
Broadly neutralizing antibodies targeting the stalk region of influenza A virus (IAV) hemagglutinin (HA) are effective in blocking virus infection both in vitro and in vivo. The highly conserved epitopes recognized by these antibodies are critical for the membrane fusion
Christoph Steininger et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 36(5), 567-574 (2003-02-21)
Twenty-one patients aged 4-78 years with influenza A virus-associated acute encephalopathy were studied. Influenza A virus could be detected only in a cerebrospinal fluid (CSF) specimen obtained from 1 of 18 patients, despite the use of a highly sensitive polymerase
Effectiveness of reverse transcription-PCR, virus isolation, and enzyme-linked immunosorbent assay for diagnosis of influenza A virus infection in different age groups.
Christoph Steininger, Michael Kundi, Stephan W Aberle, Judith H Aberle, Theresia Popow-Kraupp
Journal of Clinical Microbiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service