Skip to Content
Merck
All Photos(1)

Key Documents

LIF2010

Sigma-Aldrich

Leukemia Inhibitory Factor from mouse

10 µg, mouse recombinant LIF protein, expressed in E. coli, suitable for stem cell culture

Synonym(s):

LIF, Differentiation-stimulating Factor, D Factor, Mouse LIF, Mouse Leukemia Inhibitory Factor, Murine LIF, mLIF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

biological source

mouse

Quality Level

Assay

>95% (HPLC and SDS-PAGE)

form

liquid

specific activity

≥1 x 10(E8) U/mg

manufacturer/tradename

Chemicon®

concentration

10 μg/mL

technique(s)

cell culture | stem cell: suitable

impurities

<0.1 ng/mg Endotoxin (of LIF)

input

sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Application

RECOMMENDED QC PROTOCOL

M1 Bioassay

1. The M1 bioassay is performed using in vitro semi-solid agar cultures, which contain approximately 100 cells in 1 mL volumes of DME containing 20 % FCS in 0.3% agar.

2. Add 100 μL of sample or mLIF (10(E4) units/mL in 5% FCS in isotonic saline) in two-fold serial dilutions in duplicate to 35 mm petri dishes.

3. Add 100 μL of 5% FCS in isotonic saline to two control slides.

4. Incubate at 37°C in fully humidified atmosphere of 10% CO2 in air for 7 days.

5. Score the number of colonies that show differentiation (note: 50 units is defined as the amount of activity which results in 50% of the colonies being differentiated).

Visit www.esgro-lif.com for additional information

Manufactured by CHEMICON International, Inc. LIF is protected under US Patent nos. 5,187,077, 5,427,925, 5,443,825, 5,750,654 and 6,261,548, European Patent no. 0285 448 and related foreign patents and is not available for resale.

Unit Definition

Specific Activity: where 50 units is defined as the amount of mouse LIF required to induce differentiation in 50% of the M1 colonies in 1 mL agar cultures.

Analysis Note

Specific Activity: The activity of mouse LIF is determined by the ability to induce differentiation of murine M1 myeloid leukemic cells.The minimum detectable concentration of mouse LIF in this assay is 0.5 ng/mL
Tested negative in both aseptic and microplasmic tests.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kay Jüngling et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(14), 2100-2102 (2003-09-23)
The pluripotency and high proliferative capacity of embryonic stem (ES) cells (1-3) makes them an attractive source of different cell types for biomedical research and cell replacement therapies. A major prerequisite for these applications is the availability of a homogeneous
Megan J Ricard et al.
The Journal of biological chemistry, 288(40), 28801-28813 (2013-08-16)
The ability to differentiate embryonic stem cells (ESCs) into specific cell types is critical for improved regenerative medicine strategies, cancer chemotherapeutic approaches, and regimens to combat chronic diseases associated with aging. Subclasses of motor neurons (MNs) are generated at different
Emma C Walker et al.
The Journal of biological chemistry, 291(41), 21703-21716 (2016-08-20)
Oncostatin M (OSM) and leukemia inhibitory factor (LIF) are IL-6 family members with a wide range of biological functions. Human OSM (hOSM) and murine LIF (mLIF) act in mouse cells via a LIF receptor (LIFR)-glycoprotein 130 (gp130) heterodimer. In contrast
Eunhye Kim et al.
PloS one, 11(7), e0160289-e0160289 (2016-07-30)
Pigs provide outstanding models of human genetic diseases due to their striking similarities with human anatomy, physiology and genetics. Although transgenic pigs have been produced using genetically modified somatic cells and nuclear transfer (SCNT), the cloning efficiency was extremely low.
Hsiao-Wen Lin et al.
Journal of neuroinflammation, 6, 7-7 (2009-03-10)
Ciliary neurotrophic factor (CNTF) has been regarded as a potent trophic factor for motor neurons. However, recent studies have shown that CNTF exerts effects on glial cells as well as neurons. For instance, CNTF stimulates astrocytes to secrete FGF-2 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service