HX0640
Hydrogen peroxide solution
30%, liquid
Synonym(s):
Hydrogen peroxide solution, Hydrogen Dioxide
About This Item
Recommended Products
Product Name
Hydrogen Peroxide Solution 30%, liquid
Agency
suitable for SM 2310
suitable for SM 2320
Quality Level
vapor pressure
23.3 mmHg ( 30 °C)
form
liquid
shelf life
24 mo. (from mfg date)
concentration
29.0-32.0% (hydrogen peroxide)
impurities
≤0.001% Ammonium (NH4)
≤0.001% Chloride (Cl)
≤0.001% Nitrate (NO3)
≤0.003% Free acid (as H2SO4)
≤0.02% Phosphate (PO4)
≤0.5 ppm Iron (Fe)
≤1 ppm Heavy metals (as Pb)
≤5 ppm Sulfate (SO4)
evapn. residue
≤0.05%
color
APHA: ≤20
shipped in
ambient
storage temp.
no temp limit
SMILES string
OO
InChI
1S/H2O2/c1-2/h1-2H
InChI key
MHAJPDPJQMAIIY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Green-synthesized zinc oxide nanoparticles by Enterobacter sp.: unveiling characterization, antimicrobial potency, and alleviation of copper stress in Vicia faba (L.) plants.: This study explores the green synthesis of zinc oxide nanoparticles using Enterobacter sp., emphasizing their antimicrobial properties and ability to alleviate copper stress in plants. Hydrogen peroxide is discussed as a critical agent in the nanoparticle synthesis and stress mitigation process (Elsilk et al., 2024).
- Use of Raman spectroscopy and PCA for quality evaluation and out-of-specification identification in biopharmaceutical products.: This research utilizes Raman spectroscopy and Principal Component Analysis (PCA) to evaluate the quality of biopharmaceutical products. The study discusses the detection of hydrogen peroxide as a degradation product, crucial for ensuring product quality and compliance (Massei et al., 2024).
Analysis Note
Ammonium (NH4): 0.001% max
Assay: 29.0-32.0%
Chloride (Cl): 0.001% max
Color (APHA): 20 max
Evaporation residue: 0.05% max
Form: Clear liquid
Free acid (as H2SO4): 0.003% max
Heavy metals (as Pb): 1ppm max
Iron (Fe): 0.5ppm max
Nitrate (NO3): 0.001% max
Phosphate (PO4): 0.02% max
Sulfate (SO4): 5ppm max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service