Skip to Content
Merck
All Photos(1)

Key Documents

HX0627

Supelco

Hydrofluoric Acid

OmniTrace

Synonym(s):

Hydrogen Fluoride, Fluoric Acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

grade

for inorganic trace analysis

Quality Level

form

liquid

concentration

≥50-70% in hydrofluoric acid

pH

(at 20 °C (68 °F)strongly acid)

bp

106 °C/1013 hPa

mp

-35 °C

solubility

soluble (Water: Soluble)

density

1.16 g/cm3 at 20 °C (68 °F)

shipped in

ambient

storage temp.

room temp

SMILES string

F

InChI

InChI=1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Application

OmniTrace

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service