Skip to Content
Merck
All Photos(1)

Key Documents

BAPP1-150K

Millipore

MILLIPLEX® Bovine Acute Phase Protein Panel 1

Synonym(s):

Bovine multiplex kit, Luminex bovine immunoassay, Millipore bovine panel, bovine acute phase multiplex kit, Luminex bovine acute phase multiplex kit, bovine multiplexfibrinogen, bovine multiplex haptoglobin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

bovine

Quality Level

manufacturer/tradename

Milliplex®

parameter

25 μL sample volume (per well of a 1:100,000 dilution of serum or plasma, or a 1:50,000 dilution of milk)

technique(s)

multiplexing: suitable

input

sample type plasma
sample type serum
sample type milk

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

Acute phase proteins (APPs) constitute an integral part of the innate immune response, exhibiting significant alterations in their serum/plasma concentrations in response to inflammation, infection, or trauma. This group of proteins includes haptoglobin (Hp) and Fibrinogen, which play pivotal roles in immune response, and are frequently studied as biomarkers of systemic inflammation and disease.

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

MILLIPLEX® Bovine Acute Phase Protein Panel 1 is a 2-plex kit to be used for the simultaneous quantification of any or all of the following analytes in bovine serum, plasma and milk samples: Fibrinogen and Haptoglobin.

Features and Benefits

Configurable

Packaging

each kit sufficient for 96 wells

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service