Skip to Content
Merck
All Photos(6)

Key Documents

ABE2075

Sigma-Aldrich

Anti-TERT Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Telomerase reverse transcriptase, hEST2, hTRT, Telomerase-associated protein 2, Telomerase catalytic subunit, TP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TERT(7015)

General description

Telomerase reverse transcriptase (EC 2.7.7.49; UniProt O14746; also known as hEST2, hTRT, Telomerase-associated protein 2, Telomerase catalytic subunit, TP2) is encoded by the TERT (also known as CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TRT) gene (Gene ID 7015) in human. Telomerase reverse transcriptase (TERT) is the catalytic subunit of the telomerase responsible for adding TTAGGG repeats to the chromosome telomere ends. Cells with low or no telomerase expression lose telomere repeats during cell division, eventually resulting in cellular senescence. Most cancer cells, germ cells and embryonic stem cells express high levels of telomerase, thus contributing to pluripotency and immortality. In addition to its telomere maintenance function, telomerase also has a pro-survival role in cellular resistance against DNA damage and ensuing apoptosis. Most cancer cells are highly proliferative and express high levels of nuclear telomerase activity. Studies show that TERT shuttles from the nucleus into mitochondria upon oxidative stress induction in cancer cells. TERT mitochondrial localization helps prevent nuclear DNA damage by decreasing mitochondrial reactive oxygen species (ROS), accounting for high stress resistance especially among the cancer stem cells (CSCs) population. Following exposure to H2O2 or gamma-irradiation, cancer cells capable of excluding TERT from the nucleus display little or no DNA damage, while TERT nuclear retainment results in high DNA damage.

Specificity

This polyclonal antibody targets a sequence toward the end of the C-terminal extension (CTE) region present in spliced isoforms 1 and 3, but not in isoforms 2 and 4 of hTERT reported by UniProt (O14746).

Immunogen

Epitope: C-terminal extension (CTE).
KLH-conjugated linear peptide corresponding to a sequence from the C-terminal extension (CTE) region of human TERT.

Application

Immunocytochemistry Analysis: An 1:200 dilution from a representative lot detected TERT in A431, HeLa, HUVEC and NIH/3T3 cells.

Immunohistochemistry Analysis: An 1:250 dilution from a representative lot detected TERT in human cerebral cortex, human pancreas, and rat testis tissue sections.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-TERT Antibody is validated for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting for the detection of TERT.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected TERT in 10 µg of HeLa cell lysate.

Target description

~150 kDa observed. 127.0/120.0 kDa (human isoform 1/3) calculated.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinrui Zhao et al.
Redox biology, 20, 427-441 (2018-11-13)
Bromodomain PHD finger transcription factor (BPTF), a core subunit of nucleosome-remodeling factor (NURF) complex, plays an important role in chromatin remodeling. However, its precise function and molecular mechanism involved in hepatocellular carcinoma (HCC) growth are still poorly defined. Here, we
Balkan Sahin et al.
Cureus, 13(5), e15342-e15342 (2021-07-09)
Over 200 human telomerase reverse transcriptase (hTERT) polymorphism combinations have been implicated in the development of cancer. This study aimed to evaluate hTERT mutations in meningioma tissue and its association with meningioma. A total of 90 patients who underwent surgery
Qian Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(3), 4178-4188 (2020-01-18)
Telomerase plays a pivotal role in tumorigenesis by maintaining telomere homeostasis, a hallmark of cancer. However, the mechanisms by which telomerase is reactivated or upregulated during tumorigenesis remain incompletely understood. Here, we report that the Hippo pathway effector Yes-associated protein
Sophie Bartsch et al.
Cancers, 13(16) (2021-08-28)
The human telomerase is a key factor during tumorigenesis in prostate cancer (PCa). The androgen receptor (AR) is a key drug target controlling PCa growth and regulates hTERT expression, but is described to either inhibit or to activate. Here, we
Chaoliang Diao et al.
Molecular oncology, 15(4), 1180-1202 (2020-12-12)
Human telomerase reverse transcriptase (hTERT) plays an extremely important role in cancer initiation and development, including colorectal cancer (CRC). However, the precise upstream regulatory mechanisms of hTERT in different cancer types remain poorly understood. Here, we uncovered the candidate transcriptional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service