Skip to Content
Merck
All Photos(1)

Key Documents

AB3258

Sigma-Aldrich

Anti-Puromycin Sensitive Aminopeptidase Antibody

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NPEPPS(9520)

Specificity

Recognizes puromycin sensitive aminopeptidase.

SPECIES REACTIVITIES: It is expected that the antibody may react with other species. Reactivity with other species has not been confirmed.

Immunogen

Rat puromycin sensitive aminopeptidase.

Application

Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
This Anti-Puromycin Sensitive Aminopeptidase Antibody is validated for use in WB, IC for the detection of Puromycin Sensitive Aminopeptidase.
Western blot: 1:1,000

Immunocytochemistry: 1:1,000

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in phosphate buffered saline.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Gi Lee et al.
Bioscience, biotechnology, and biochemistry, 79(9), 1414-1421 (2015-04-30)
A gel-free and label-free quantitative proteomic approach based on a spectral counting strategy was performed to discover prolificacy-related proteins. Soluble proteins of porcine placenta from small litter size group (SLSG) and large litter size group (LLSG) were extracted and subsequently
Helena R Zimmermann et al.
Journal of neurochemistry, 146(2), 133-144 (2018-01-18)
Characterization of the molecular signaling pathways underlying protein synthesis-dependent forms of synaptic plasticity, such as late long-term potentiation (L-LTP), can provide insights not only into memory expression/maintenance under physiological conditions but also potential mechanisms associated with the pathogenesis of memory
Wenzhong Yang et al.
Molecular psychiatry, 26(6), 1880-1897 (2020-05-06)
The AMP-activated protein kinase (AMPK) is a molecular sensor to maintain energy homeostasis. The two isoforms of the AMPK catalytic subunit (AMPKα1 and α2) are both expressed in brains, but their roles in cognition are unknown. We generated conditional knockout
Helena R Zimmermann et al.
The Journal of clinical investigation, 130(7), 3511-3527 (2020-03-28)
AMPK is a key regulator at the molecular level for maintaining energy metabolism homeostasis. Mammalian AMPK is a heterotrimeric complex, and its catalytic α subunit exists in 2 isoforms: AMPKα1 and AMPKα2. Recent studies suggest a role of AMPKα overactivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service