Skip to Content
Merck
All Photos(1)

Key Documents

8.52187

Sigma-Aldrich

Fmoc-Ser(tBu)-Ser(psiMe,Mepro)-OH

≥97.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Ser(tBu)-Ser(psiMe,Mepro)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34N2O7
Molecular Weight:
510.58
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Ser(tBu)-Ser(psiMe,Mepro)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥97% (TLC)
≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-8°C

General description

This pseudoproline dipeptide is undoubtedly the most effective and simplest to use tool for overcoming aggregation and enhancing peptide quality in Fmoc SPPS for peptide containing the Ser-Ser dipeptide motif. It consists of a dipeptide in which the serine residue has been reversibly protected as a structure-breaking proline-like TFA-labile oxazolidine. The reason the pseudoproline residue is introduced as a dipeptide is because it avoids the need to acylate the hindered oxazolidine nitrogen. This has the added advantage of extending the peptide chain by two residues in one step. The serine residue is regenerated during the normal TFA-mediated cleavage reaction.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS

Linkage

Replaces: 05-20-1012

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in CH₂Cl₂): -21.5 - -17.5 °
Purity (TLC(157A)): ≥ 97 %
Purity (TLC(CMA2)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Single impurities (HPLC, area%): ≤ 1.20 % (a/a)
Acetate (IC): ≤ 0.1 %
Ethyl acetate (HS-GC): ≤ 2.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service