Skip to Content
Merck
All Photos(1)

Key Documents

8.52084

Sigma-Aldrich

Fmoc-Dab(ivDde)-OH

Novabiochem®

Synonym(s):

Fmoc-Dab(ivDde)-OH, N-α-Fmoc-N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)-3-methylbutyl-L-diaminobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H38N2O6
CAS Number:
Molecular Weight:
546.65
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

InChI

1S/C32H38N2O6/c1-19(2)15-26(29-27(35)16-32(3,4)17-28(29)36)33-14-13-25(30(37)38)34-31(39)40-18-24-22-11-7-5-9-20(22)21-10-6-8-12-23(21)24/h5-12,19,24-25,35H,13-18H2,1-4H3,(H,34,39)(H,37,38)/t25-/m0/s1

InChI key

IMMMALLSYWTSAX-VWLOTQADSA-N

General description

This orthogonally-protected diaminobutyric acid derivative is based on the hindered Dde variant ivDde. The side-chain ivDde group is considerably more stable to piperidine than Dde and is less prone to migrate from protected to unprotected side-chains [1] and from side-chain to γ-amino group [2].When removing ivDde in the presence of allyl-based protecting groups, allyl alcohol should be included in the deprotection solution to prevent reduction of the allyl group [3].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] S. R. Chhabra, et al. (1998) Tetrahedron Lett., 39, 1603.
[2] R. Wilhelm, personal communication.
[3] B. Rohwedder, et al. (1998) Tetrahedron Lett., 39, 1175.
[4] S. K. Sharma, et al. (1999) J. Peptide Res., 53, 501.

Linkage

Replaces: 04-12-1196

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(157A)): ≥ 98 %
Purity (TLC(157B)): ≥ 98 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service