Skip to Content
Merck
All Photos(1)

Key Documents

8.18702

Sigma-Aldrich

Bromoform

(stabilised with 2-methyl-2-butene) for synthesis

Synonym(s):

Bromoform, Tribromomethane, Methane tribromide, Methyl tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-854-6
NACRES:
NA.22

vapor pressure

7.5 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

potency

933 mg/kg LD50, oral (Rat)

bp

149.5 °C/1013 mbar

mp

9.2 °C

solubility

3.2 g/L

density

2.89 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/CHBr3/c2-1(3)4/h1H

InChI key

DIKBFYAXUHHXCS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area %): ≥ 98.0 %
Identity (IR-spectrum): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service