Skip to Content
Merck
All Photos(1)

Key Documents

8.18591

Sigma-Aldrich

Thiourea

for synthesis

Synonym(s):

Thiourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4N2S
CAS Number:
Molecular Weight:
76.12
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-543-5
NACRES:
NA.22

Quality Level

form

solid

autoignition temp.

440 °C (Dust)

potency

1750 mg/kg LD50, oral (Rat)
>2800 mg/kg LD50, skin (Rabbit)

reaction suitability

reagent type: reductant

pH

6-8 (20 °C, 50 g/L in H2O)

mp

169-173 °C

solubility

137 g/L

density

1.405 g/cm3 at 20 °C

bulk density

640 kg/m3

storage temp.

2-30°C

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Analysis Note

Assay (ex N): ≥ 98.0 %
Assay (Fe): ≤ 0.001 %
Identity (IR): passes test

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service