Skip to Content
Merck
All Photos(1)

Documents

579051

Sigma-Aldrich

TAPI-1

≥97% (HPLC), solid, MMP, TACE inhibitor, Calbiochem®

Synonym(s):

TAPI-1, N-(R)-[2-(Hydroxyaminocarbonyl)methyl]-4-methylpentanoyl-L-naphthylalanyl-L-alanine, 2-aminoethyl Amide, TNF-α Protease Inhibitor-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37N5O5
CAS Number:
Molecular Weight:
499.60
UNSPSC Code:
12352200
NACRES:
NA.77

product name

TAPI-1, TAPI-1, CAS 171235-71-5, is a structural analog of TAPI-0 with similar in vitro efficacy for the inhibition of MMPs and TACE. Blocks the shedding of several cell surface proteins.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

methanol: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C26H37N5O5/c1-16(2)12-21(15-23(32)31-36)25(34)30-22(26(35)29-17(3)24(33)28-11-10-27)14-18-8-9-19-6-4-5-7-20(19)13-18/h4-9,13,16-17,21-22,36H,10-12,14-15,27H2,1-3H3,(H,28,33)(H,29,35)(H,30,34)(H,31,32)/t17-,21?,22-/m0/s1

InChI key

AWNBSWDIOCXWJW-OWHMDLSXSA-N

General description

A structural analog of TAPI-0 (Cat. No. 579050) with similar in vitro efficacy for the inhibition of MMPs and TACE (TNF-α converting enzyme/ADAM17). TAPI-1 also blocks the shedding of several cell surface proteins such as IL-6 receptor, p60 TNF receptor, and p80 TNF receptor. Blocks constitutive (IC50 = 8.09 µM) and muscarinic receptor-stimulated (IC50 = 3.61 µM) sAAPα release in HEK 293 cells expressing M3 muscarinic receptors. A 10 mM (500 µg/100 µl) solution of TAPI-1 (Cat. No. 579050) in DMSO is also available.
A structural analog of TAPI-0 (Cat. No. 579050) with similar in vitro efficacy for the inhibition of MMPs and TACE (TNF-α convertase; ADAM17). However, TAPI-1 is more stable in serum than TAPI-0. TAPI-1 Blocks the shedding of several cell surface proteins such as TNF-α, IL-6 receptor, and p60 and p80 TNF receptors. Inhibits constitutive (IC50 = 8.09 µM) and muscarinic receptor-stimulated (IC50 = 3.61 µM) sAPPa release in HEK-293 cells expressing muscarinic receptors. Also blocks the TACE-dependent constitutive release of co-transfected APP(695) (IC50 = 920 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMPs and TACE
Product does not compete with ATP.
Reversible: no
Target IC50: 8.09 µM and 3.61 µM blocking constitutive and muscarinic receptor-stimulated sAAPα release in HEK 293 cells expressing M3 muscarinic receptors, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Slack, B.E., et al. 2001. Biochem. J.357, 787.
Vincent, B., et al. 2001. J. Biol. Chem.276, 37743.
Hooper, N.M., et al. 1997. Biochem. J.321, 265.
Crowe, P.D., et al. 1995. J. Exp. Med.181, 1205.
Mullberg, J., et al. 1995. J. Immunol.155, 5198.
Mohler, K.M., et al. 1994. Nature370, 218.

Legal Information

Manufactured exclusively by Peptides International, Louisville, Kentucky, USA.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Liu et al.
PloS one, 15(7), e0236175-e0236175 (2020-07-23)
Adenoviruses cause upper respiratory infections, conjunctivitis, keratitis, and gastrointestinal illness. These can be fatal in immunocompromised individuals. Adenoviruses have also been engineered into viral vectors to deliver therapeutic genes or induce immunity as vaccine carriers. The success of ocular gene
Bradley R Kraemer et al.
Neuroscience, 453, 222-236 (2020-12-01)
The p75 neurotrophin receptor (p75NTR) is a multifunctional protein that regulates cellular responses to pathological conditions in specific regions of the nervous system. Activation of p75NTR in certain neuronal populations induces proteolytic processing of the receptor, thereby generating p75NTR fragments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service