Skip to Content
Merck
All Photos(1)

Documents

5.31131

Sigma-Aldrich

USP1-UAF1 Inhibitor, ML323

Synonym(s):

USP1-UAF1 Inhibitor, ML323, N-(4-(1H-1,2,3-Triazol-1-yl)benzyl)-2-(2-isopropylphenyl)-5-methylpyrimidin-4-amine, USP1–UAF1 Deubiquitinase complex Inhibitor, ML 323

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24N6
CAS Number:
Molecular Weight:
384.48
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CC1=CN=C(N=C1NCC2=CC=C(C=C2)N3C=CN=N3)C4=CC=CC=C4C(C)C

General description

A cell-permeable, non-toxic triazolylbenzyl-pyrimidinamine compound that acts as a highly selective, potent, and reversible inhibitor of ubiquitin-specific protease 1 (USP1)-USP1 associated factor 1 (UAF1) deubiquitinase (DUB) (IC50 = 76, 174, and 820 nM in Ub-Rhodamine; K63 linked diubiquitin; and Ub-PCNA substrate based assays) without disrupting USP1-UAF1 association. The inhibition appears to be allosteric in nature with inhibition constants of 68 nM and 183 nM (for free enzyme and enzyme-substrate complex, respectively). Displays excellent selectivity over 18-DUBs including USP1 and USP46-UAF1, related deSUMOylases & deneddylases, 70 unrelated proteases, and 451 kinases. Effectively sensitizes cisplatin-resistant NSCLC H596 cells and U2OS osteosarcoma cells to cisplatin (Cat. No. 232120) and blocks cell cycle at the S phase (EC50 = 171 and 59 nM for 1:1 and 1:4 ratio of cisplatin to ML323 in H596 cells).
A cell-permeable, non-toxic triazolylbenzyl-pyrimidinamine compound that acts as a highly selective, potent, and reversible inhibitor of ubiquitin-specific protease 1 (USP1)-USP1 associated factor 1 (UAF1) deubiquitinase (DUB) (IC50 = 76, 174, and 820 nM in Ub-Rhodamine; K63 linked diubiquitin; and Ub-PCNA substrate based assays) without disrupting USP1-UAF1 association. The inhibition appears to be allosteric in nature with inhibition constants of 68 nM and 183 nM (for free enzyme and enzyme-substrate complex, respectively). Displays excellent selectivity over 18-DUBs including USP1 and USP46-UAF1, related deSUMOylases & deneddylases, 70 unrelated proteases, and 451 kinases. Effectively sensitizes cisplatin-resistant NSCLC H596 cells and U2OS osteosarcoma cells to cisplatin (Cat. No. 232120) and blocks cell cycle at the S phase (EC50 = 171 and 59 nM for 1:1 and 1:4 ratio of cisplatin to ML323 in H596 cells).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
USP1-UAF1 deubiquitinase (DUBs) complex
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Dexheimer, T.S., et al. 2014. J. Med. Chem.57, in press.
Liang, Q., et al. 2014. Nat Chem Biol.10, 298.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service