Skip to Content
Merck
All Photos(1)

Key Documents

3570

Millipore

Glycine

Synonym(s):

Glycine, Gly, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.54

Assay

>99% (by assay)

Quality Level

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 CFU/g Total microorganism
≤6.0 EU/g Endotoxin

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

water: 250 mg/mL

cation traces

Fe: ≤10 ppm
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A non-essential, optically inactive, non-chiral amino acid. It is the smallest of the 20 natural amino acids. Acts as an inhibitory neurotransmitter, particularly in brain stem and medulla. Shown to block NMDA-stimulated dopamine release in fetal mesencephalic cell cultures. Also reported to increase NMDA-receptor mediated synaptic potential in cortical neurons.
A nonpolar, uncharged amino acid.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Warning

Toxicity: Standard Handling (A)

Other Notes

Shapiro, S. 1997. Neurosurgery 40, 168.
Mount, H., et al. 1991. Mol. Pharmacol.39, 205.
Thompson, A.M., et al. 1989. Nature338, 422.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service