Skip to Content
Merck
All Photos(1)

Key Documents

1.93411

Supelco

Citric acid monohydrate

EMPLURA®

Synonym(s):

Citric acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
4018641
MDL number:
UNSPSC Code:
12352106

product line

EMPLURA®

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

storage temp.

room temp

SMILES string

OC(CC(O)(C(O)=O)CC(O)=O)=O.O

InChI

1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2

InChI key

YASYEJJMZJALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93411.0521: 500G, Minimum order quantity: 20
1.93411.5021: 5 kg, Minimum order quantity: 4
1.93411.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

345.0 °F - closed cup

Flash Point(C)

173.9 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service