Skip to Content
Merck
All Photos(1)

Key Documents

1.93002

Supelco

Acetic acid glacial 100%

for analysis EMPARTA® ACS

Synonym(s):

Acetic acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106

grade

ACS reagent

vapor density

2.07 (vs air)

product line

EMPARTA®

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

20-25°C

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93002.0521: 500ML, Minimum order quantity: 20
1.93002.2521: 2.5L, Minimum order quantity: 4
1.93002.9026: 25 L, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service