Skip to Content
Merck
All Photos(1)

Documents

1.46694

Millipore

ReadyTube® 10 RVS Broth

Suitable for the selective enrichment of Salmonella, pkg of (10 ml in 17 ml tube with blue cap (20 tubes per box))

Synonym(s):

Rappaport Vassiliadis -Soya Broth, Rappaport Vassiliadis Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214
NACRES:
NA.74

Agency

ISO 19250
ISO 6579
ISO 6785 I IDF 93
ISO FDIS 6579-1

Quality Level

sterility

sterile; autoclaved

form

liquid (ready-to-use)

feature

ready-to-use

packaging

pkg of (10 ml in 17 ml tube with blue cap (20 tubes per box))

pH

5.2 ( in H2O)

application(s)

bioburden testing
environmental
food and beverages

storage temp.

2-25°C

suitability

selective for Salmonella spp.

General description

ReadyTube® 10 RVS Broth is a modification of the original Rappaport medium with soya peptone instead of tryptone (peptone from casein) to improve recovery rates of Salmonella. The efficiency of RVS broth is based on: (a) the ability of Salmonella spp. to multiply at relatively high osmotic pressures, at relatively low pH values, at a high temperature, and with modest nutritional requirements; and (b) the suppression of the toxic effect of malachite green towards Salmonellae by the presence of magnesium chloride.

Application

ReadyTube® 10 RVS Broth is suitable for the selective enrichment of Salmonella from food and animal feed, water, and other materials.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Packaging

10 ml in 17 ml tube with blue cap (20 tubes per box)

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service