Skip to Content
Merck
All Photos(1)

Documents

14-306-M

Sigma-Aldrich

GSK3β Protein, active, 10 µg

Active, N-terminal His6-tagged recombinant human GSK3β with an H350L mutation, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.26

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

form

liquid

mol wt

Mw 51 kDa

composition

mercaptoethanol, 0.1-1%
saccharose, 5-10%

manufacturer/tradename

Upstate®

storage condition

(Tightly closed)

technique(s)

activity assay: suitable (kinase)

solubility

water: soluble

NCBI accession no.

UniProt accession no.

Gene Information

human ... GSK3B(2932)

General description

Glycogen synthase kinase 3β (GSK3β) is a serine-threonine kinase that is present in most eukaryotes. The GSK3β gene is located on the human chromosome at 3q13.3. GSK3β is made up of an N-terminal β-sheet domain that is coupled to the C-terminal α-helical domain.

Research area: Cell Signaling

Biochem/physiol Actions

Glycogen synthase kinase 3β (GSK3β) regulates glycogen metabolism via an insulin-dependent pathway. It participates in many signaling pathways including cellular processes such as cell cycle, cell proliferation, and inflammation. GSK3β mediated-negative regulation of insulin signaling pathway leads to type 2 diabetes. GSK3β functions as a tumor suppressor that represses neoplastic transformation in cells. It is an essential regulator of the Wnt/β-catenin signaling pathway. Abnormal regulation of GSK3β is associated with cardiovascular disease, bipolar disorder, and neurodegenerative diseases.

Packaging

Also available in 250μg size (2x125μg)--call for pricing and availability and reference catalog number 14-306M when ordering the 250μg size.

Quality

Routinely evaluated by phosphorylation of Phospho-Glycogen Synthase Peptide-2

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

antibody

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

rystal structure of glycogen synthase kinase 3 beta: structural basis for phosphate-primed substrate specificity and autoinhibition
Dajani R, et al.
Cell, 105(6), 721-732 (2009)
S Phukan et al.
British journal of pharmacology, 160(1), 1-19 (2010-03-25)
Glycogen synthase kinase-3 beta (GSK3beta) is a multifunctional serine/threonine kinase which was originally identified as a regulator of glycogen metabolism. It plays a key role in the regulation of numerous signalling pathways including cellular process such as cell cycle, inflammation
Jia Luo
Cancer letters, 273(2), 194-200 (2008-07-09)
Glycogen synthase kinase 3beta (GSK3beta), a multifunctional serine/threonine kinase found in all eukaryotes, had been initially identified as a key regulator of insulin-dependent glycogen synthesis. It is now known that GSK3beta functions in diverse cellular processes including proliferation, differentiation, motility
The alpha-isoform of glycogen synthase kinase-3 from rabbit skeletal muscle is inactivated by p70 S6 kinase or MAP kinase-activated protein kinase-1 in vitro.
Sutherland, C and Cohen, P
Febs Letters, 338, 37-42 (1994)
Ethanol impairs insulin-stimulated neuronal survival in the developing brain: role of PTEN phosphatase
Xu, J., et al
The Journal of Biological Chemistry, 278, 26929-26937 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service