Skip to Content
Merck
All Photos(1)

Key Documents

126674

Sigma-Aldrich

Albumin, Mouse

Synonym(s):

Albumin, Mouse, MSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 8468

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

ELISA: suitable

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Plasma protein that is useful as a stabilizing agent and a carrier protein.
Plasma protein that is useful as a stabilizing agent and as a carrier protein.

Application

Albumin, mouse has been used as a component of PBS for in vivo examination of (E, E)-1-fluoro-2,5-bis(3-hydroxycarbonyl-4-hydroxy)styrylbenzene (FSB) and yellow fluorescent protein (YFP) positive cells in mice.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arieke Suzanna Berendina Kampstra et al.
Annals of the rheumatic diseases, 78(7), 908-916 (2019-06-04)
Autoantibodies against post-translationally modified proteins (anti-modified protein antibodies or AMPAs) are a hallmark of rheumatoid arthritis (RA). A variety of classes of AMPAs against different modifications on proteins, such as citrullination, carbamylation and acetylation, have now been described in RA.
Petar Marinković et al.
Brain : a journal of neurology, 142(4), 1051-1062 (2019-03-09)
Pathological alterations of tau protein play a significant role in the emergence and progression of neurodegenerative disorders. Tauopathies are characterized by detachment of the tau protein from neuronal microtubules, and its subsequent aberrant hyperphosphorylation, aggregation and cellular distribution. The exact
Christian Schön et al.
PloS one, 7(12), e53547-e53547 (2013-01-10)
Tauopathies are widespread neurodegenerative disorders characterised by the intracellular accumulation of hyperphosphorylated tau. Especially in Alzheimer's disease, pathological alterations in the retina are discussed as potential biomarkers to improve early diagnosis of the disease. Using mice expressing human mutant P301S

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service