Skip to Content
Merck
All Photos(1)

Key Documents

1.16978

Supelco

Phosphate Test Strips, reflectometric

5-120 mg/L (PO₄³⁻), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Phosphate chemical test strips, Phosphate test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Phosphate Test, reflectometric, 5-120 mg/L (PO43-), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

phosphate

measuring range

5-120 mg/L (PO43-)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

15-25°C

General description

Method: reflectometric with test strips and reagent 5 - 120 mg/l PO₄³⁻ Reflectoquant®
The Reflectoquant Phosphate test includes test strips and all necessary additional reagents. It is suitable for determining orthophosphate in wastewater, soil samples, fertilizers and food. In sulfuric solution orthophosphate ions (PO43-) react with molybdate ions to form molybdophosphoric acid. This is reduced to phosphomolybdenum blue (PMB) that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the phosphate concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for phosphate. Further quantitative determination methods can be found with our Spectroquant tests.

Application


  • Environmental Monitoring: Phosphate tests are extensively used in environmental monitoring to assess the levels of phosphate in water bodies. These tests help in evaluating the impact of agricultural runoff, industrial discharge, and wastewater treatment on water quality. Recent studies have focused on using selective colorimetric nano-sensing solutions to determine phosphate ions in drinking water samples, highlighting their importance in ensuring safe and clean water supplies (Salem and Draz, International Journal of Environmental Analytical Chemistry, 2020).

Analysis Note

function control: conforms
Assay
- standard 1:
8 - 12 mg/l
- standard 2: 45 - 55 mg/l
- standard 3: 90 - 110 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service