Skip to Content
Merck
All Photos(1)

Key Documents

1.14558

Supelco

Ammonium Test, photometric

photometric, 0.20-8.00 mg/L (NH4-N), 0.26-10.30 mg/L (NH4+), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Ammonium Cell Test, photometric, 0.20-8.00 mg/L (NH4-N), 0.26-10.30 mg/L (NH4+), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

ammonium

measuring range

0.20-8.00 mg/L (NH4-N)
0.26-10.30 mg/L (NH4+)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Indophenol blue)

storage temp.

15-25°C

General description

Method: Photometric, 0.20 - 8.00 mg/L (NH4-N), 0.26 - 10.30 mg/L (NH4+), Spectroquant®

The method corresponds to ISO 23695, a norm standardizing the use of this "small-scale sealed tube method" for this important parameter. The method is also analogous to EPA 350.1, APHA 4500-NH3 F, ISO 7150-1, and DIN 38406-5.

The Spectroquant® Ammonium Cell Test allows the cyanide-free quantification of ammonium in wastewater, groundwater, surface water, and seawater. Typically, ammonium enters water sources as runoff from fertilized agricultural areas and enhanced levels are important indicators for faecal pollution.Further fields of applications are the analysis of foods & beverages and of environmental and agricultural samples (e.g. soil).

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service