Skip to Content
Merck
All Photos(1)

Key Documents

1.00289

Millipore

L-Glutamine

for biochemistry

Synonym(s):

L-Glutamic acid-5-amide, Gln, (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723797
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-292-1
NACRES:
NA.21

Quality Level

form

solid

potency

7500 mg/kg LD50, oral (Rat)

pH

4.0-6.0 (20 °C, 25 g/L in H2O)

mp

185 °C (dec.) (lit.)

solubility

100 g/L

density

1.47 g/cm3 at 20 °C

bulk density

640 kg/m3

storage temp.

2-30°C

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Low-protein diet supplemented with 1% L-glutamine improves growth performance, serum biochemistry, redox status, plasma amino acids, and alters fecal microbiota in weaned piglets.: This study demonstrates how L-glutamine supplementation in a low-protein diet can significantly enhance growth metrics and overall health in weaned piglets (Li et al., 2024).

  • Functionalization of β-cyclodextrin metal-organic frameworks with gelatin and glutamine for drug delivery of curcumin to cancerous cells.: This article covers the development of a novel drug delivery system using glutamine-modified frameworks for targeted cancer therapy (Sadeh et al., 2024).

Biochem/physiol Actions

L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): 99.0 - 101.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine powder
Appearance of solution (25 g/l; CO₂-free water): clear and colorless
Spec. rotation (α 20/D, 100 g/l, hydrochloric acid 2 mol/l, calc. on dried substance): +31,5 to +33,0
Chloride (Cl): ≤ 200 ppm
Sulfate (SO₄): ≤ 200 ppm
Heavy metals (as Pb): ≤ 10 ppm
As (Arsenic): ≤ 5 ppm
Ca (Calcium): ≤ 10 ppm
Co (Cobalt): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
K (Potassium): ≤ 5 ppm
Mg (Magnesium): ≤ 5 ppm
Na (Sodium): ≤ 50 ppm
Zn (Zinc): ≤ 5 ppm
Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity): ≤ 0.3 %
Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.1 %
Ninhydrin-positive substances (LC)(total impurities): ≤ 0.5 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service