Skip to Content
Merck
All Photos(1)

Key Documents

07-728

Sigma-Aldrich

Anti-HRI Antibody

serum, Upstate®

Synonym(s):

Anti-HCR, Anti-HRI, Anti-LEMSPAD, Anti-hHRI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rabbit, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... EIF2AK1(27102)
mouse ... Eif2Ak1(15467)

Specificity

HRI

Immunogen

6His-tagged fusion protein corres-ponding to residues 1-138 of mouse heme-regulated inhibitor (HRI)

Application

Detect HRI using this Anti-HRI Antibody validated for use in IP & WB.

Quality

Routinely evaluated by immunoblot on lysates from mouse reticulocytes.

Target description

72kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Chen et al.
Trends in biochemical sciences, 20(3), 105-108 (1995-03-01)
Protein synthesis is regulated by the phosphorylation of the alpha-subunit of eukaryotic initiation factor 2 (eIF-2 alpha) in a variety of cells. At present, there are two distinct mammalian eIF-2 alpha kinases that have been cloned, the double-stranded-RNA-dependent eIF-2 alpha
Takahiro Sakai et al.
Antioxidants (Basel, Switzerland), 10(9) (2021-09-29)
Lung fibrosis is the primary pathology in idiopathic pulmonary fibrosis and is considered to result from an increase in reactive oxygen species (ROS) levels in alveolar epithelial cells. However, the exact mechanism underlying lung fibrosis remains unclear and there is
Visibly stressed: the role of eIF2, TIA-1, and stress granules in protein translation.
Anderson, Paul and Kedersha, Nancy
Cell Stress & Chaperones, 7, 213-221 (2002)
Maryam Rafie-Kolpin et al.
Biochemistry, 42(21), 6536-6544 (2003-05-28)
In heme deficiency, protein synthesis is inhibited by the activation of the heme-regulated eIF2alpha kinase (HRI) through its multiple autophosphorylation. Autophosphorylation sites in HRI were identified in order to investigate their functions. We found that there were eight major tryptic
Nicolò Musner et al.
ASN neuro, 8(2) (2016-04-21)
In peripheral nerves, P0 glycoprotein accounts for more than 20% of myelin protein content. P0 is synthesized by Schwann cells, processed in the endoplasmic reticulum (ER) and enters the secretory pathway. However, the mutant P0 with S63 deleted (P0S63del) accumulates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service