Skip to Content
Merck
All Photos(1)

Key Documents

H-102

Supelco

4-Hydroxyduloxetine-beta-D-glucuronide solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO8S
Molecular Weight:
489.54
MDL number:
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

4-Hydroxyduloxetine glucuronide is a metabolite of duloxetine. Duloxetine, marketed in the US under the trade name Cymbalta® is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis. This Certified Spiking Solution® is applicable for use in applications such as clinical toxicology, urine drug testing, and forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Cymbalta is a registered trademark of Eli Lilly and Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Smit et al.
Oncogene, 30(35), 3735-3744 (2011-04-12)
Anoikis (detachment-induced apoptosis) prevents the survival of cells at inappropriate sites of the body and can therefore act as a barrier to metastasis. In a function-based genome-wide screen, we have previously identified the neurotrophic tyrosine kinase receptor TrkB as a
Deyin Xing et al.
Cancer research, 69(21), 8231-8235 (2009-10-22)
Uterine leiomyosarcoma (ULMS) is a rare gynecologic malignancy with a low survival rate. Currently, there is no effective treatment for ULMS. Infrequent occurrences of human ULMS hamper the understanding of the initiation and progression of the disease, thereby limiting the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service