Skip to Content
Merck
All Photos(1)

Key Documents

D-067

Supelco

Digitoxin solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H64O13
CAS Number:
Molecular Weight:
764.94
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

CC1OC(CC(O)C1O)OC2C(O)CC(OC2C)OC3C(O)CC(OC3C)OC4CCC5(C)C(CCC6C5CCC7(C)C(CCC67O)C8=CC(=O)OC8)C4

InChI

1S/C41H64O13/c1-20-36(46)29(42)16-34(49-20)53-38-22(3)51-35(18-31(38)44)54-37-21(2)50-33(17-30(37)43)52-25-8-11-39(4)24(15-25)6-7-28-27(39)9-12-40(5)26(10-13-41(28,40)47)23-14-32(45)48-19-23/h14,20-22,24-31,33-38,42-44,46-47H,6-13,15-19H2,1-5H3

InChI key

WDJUZGPOPHTGOT-UHFFFAOYSA-N

General description

A cardiac glycoside and antiarrhythmic drug similar in structure and effect to digoxin. Digitoxin is monitored by clinical labs to ensure patients remain within the drug′s therapeutic range.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Termination of refractory focal status epilepticus by the P-glycoprotein inhibitor verapamil.
S Pirker et al.
European journal of neurology, 18(12), e151-e151 (2011-11-22)
An elderly man with atrial fibrillation and exacerbated diabetes.
Ketil Arne Espnes et al.
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 132(9), 1103-1105 (2012-05-23)
Ryan Matsuda et al.
Journal of chromatography. A, 1265, 114-122 (2012-10-25)
High-performance affinity chromatography (HPAC) was used to examine the changes in binding that occur for the sulfonylurea drug glibenclamide with human serum albumin (HSA) at various stages of glycation for HSA. Frontal analysis on columns containing normal HSA or glycated
Thomas M Beale et al.
Organic letters, 15(6), 1358-1361 (2013-03-08)
The cardiac glycoside natural product digitoxin was selectively glycosylated at one of its five hydroxyl groups using a borinic acid derived catalyst. This method provided access to the glycosylation pattern characteristic of a subclass of natural products from Digitalis purpurea.
Ioannis Prassas et al.
Molecular cancer therapeutics, 10(11), 2083-2093 (2011-08-24)
Cardiac glycosides (e.g., digoxin, digitoxin) constitute a diverse family of plant-derived sodium pump inhibitors that have been in clinical use for the treatment of heart-related diseases (congestive heart failure, atrial arrhythmia) for many years. Recently though, accumulating in vitro and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service