Skip to Content
Merck
All Photos(1)

Key Documents

D-003

Supelco

1,3-Dinitroglycerin solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2O7
CAS Number:
Molecular Weight:
182.09
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OC(CO[N+]([O-])=O)CO[N+]([O-])=O

InChI

1S/C3H6N2O7/c6-3(1-11-4(7)8)2-12-5(9)10/h3,6H,1-2H2

InChI key

ASIGVDLTBLZXNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dinitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution® is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V Hutt et al.
Arzneimittel-Forschung, 44(12), 1317-1321 (1994-12-01)
In the course of this study both the bioavailability and main pharmacokinetic parameters of the glyceryl trinitrate (GTN, CAS 55-63-0) metabolites 1,2 and 1,3-glyceryl dinitrate (1,2-GDN and 1,3-GDN) were to be determined following transdermal application of a glyceryl trinitrate test
K M Jensen et al.
Arzneimittel-Forschung, 44(8), 951-953 (1994-08-01)
The plasma profiles of nitroglycerin (glyceryl trinitrate, GTN, CAS 55-63-0) and its dinitrate metabolites (1,2-GDN and 1,3-GDN) were estimated after sublingual administration of GTN tablets 0.5 mg (Nitromex) given in single doses to 20 volunteers. Blood samples (totally 20) were
B P Booth et al.
Canadian journal of physiology and pharmacology, 71(8), 629-632 (1993-08-01)
It has previously been shown that the vasodilatory response to glyceryl trinitrate (GTN) was decreased during hypothermic cardiopulmonary bypass. The purpose of these experiments was to determine the effect of temperature on GTN-induced relaxation and on GTN biotransformation in rabbit
B P Booth et al.
Anesthesia and analgesia, 78(5), 848-856 (1994-05-01)
Glyceryl trinitrate (GTN) is used to control arterial blood pressure during cardiopulmonary bypass (CPB) procedures, but its effects are often decreased during the period of extracorporeal support. The plasma and urine concentrations of GTN and glyceryl-1,2-dinitrate (1,2-GDN) and glyceryl-1,3-dinitrate (1,3-GDN)
T Kikkoji et al.
Pharmaceutical research, 8(10), 1231-1237 (1991-10-01)
The percutaneous penetration kinetics of the antianginal, nitroglycerin (GTN), and its primary metabolites, 1,2- and 1,3-glyceryl dinitrate (1,2- and 1,3-GDN), were evaluated in vitro, using full-thickness hairless mouse skin. GTN and the 1,2- and 1,3-GDNs were applied (a) in aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service