Key Documents
Z616907
Vacuum accessories for Buchi® vacuum pumps
Needle valve gauge
About This Item
UNSPSC Code:
41104804
NACRES:
NB.43
packaging
pack of 1 ea
manufacturer/tradename
Büchi 047291
General description
The simplest way to maintain vacuum control is through this cost-effective needle valve. Air is drawn in through a fine regulation valve and the resulting vacuum is set. A manometer displays the pressure. A Woulff bottle is recommended with this valve so that it can be a buffer vessel and condensate trap.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG