Z567574
Aldrich® Benchclamp™ 3-prong
dual adjustment grippy knobs, size S
Synonym(s):
bench clamps
All Photos(2)
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
Recommended Products
size
S
jaw capacity
0-30 mm
prong W
6 mm
Related Categories
General description
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Round extension arms allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames. Supplied with vinyl and fiberglass prong covers for high temperature applications.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Round extension arms allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames. Supplied with vinyl and fiberglass prong covers for high temperature applications.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)