material
borosilicate glass
colorless cap
colorless tube
Joint
joint: ST/NS24/40
feature
graduations: no
L
125 mm
Looking for similar products? Visit Product Comparison Guide
General description
Desiccant area- 35 mm L
The Aldrich drying tubes are made of borosilicate glass and are designed with a bent goose-neck and standard taper jointed end. The tube also contains a desiccant area to hold the desiccant. Joint angle is a 75°.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service