SYNF791100
Synthware™ modified evaporation flask
capacity 100 mL, neck joint: ST/NS 14/20
Synonym(s):
Synthware F791100, rotary evaporator flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
glass flask
round bottom flask
Joint
joint: ST/NS neck14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F791100
capacity
100 mL
Related Categories
General description
Low profile design sits upright on bench and also provides for easy product access with a spatula. The unique flask shape is excellent for the controlled evaporation of foaming solvents such as toluene or xylene on a rotary evaporator
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)