Skip to Content
Merck
All Photos(1)

Documents

N27314

Sigma-Aldrich

3-Nitrotoluene

99%

Synonym(s):

1-Methyl-3-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
1906910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.73 (vs air)

vapor pressure

1 mmHg ( 50.2 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

230-231 °C (lit.)

mp

14-16 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)[N+]([O-])=O

InChI

1S/C7H7NO2/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3

InChI key

QZYHIOPPLUPUJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F

Flash Point(C)

98 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Struijs et al.
The Science of the total environment, 57, 161-170 (1986-12-01)
The biodegradation of 2-, 3- and 4-nitrotoluene was investigated in a simple laboratory test. All three isomers are shown to be biodegradable in a die-away test after adaptation of the inoculum, though different results were obtained with different types of
B E Haigler et al.
Applied and environmental microbiology, 60(9), 3466-3469 (1994-09-01)
A strain of Pseudomonas sp. was isolated from nitrobenzene-contaminated soil and groundwater on 2-nitrotoluene as the sole source of carbon, energy, and nitrogen. Bacterial cells growing on 2-nitrotoluene released nitrite into the growth medium. The isolate also grew on 3-methylcatechol
L E Hallas et al.
Applied and environmental microbiology, 45(4), 1234-1241 (1983-04-01)
The transformation of mono- and dinitroaromatic compounds was measured in sewage effluent maintained under aerobic or anaerobic conditions. Most of the nitrobenzene, 3- and 4-nitrobenzoic acids, and 3- and 4-nitrotoluenes and much of the 1,2- and 1,3-dinitrobenzenes disappeared both in
D E Rickert et al.
Chemico-biological interactions, 52(2), 131-139 (1984-12-01)
The mononitrotoluenes are important industrial chemicals which display isomeric specificity in their ability to induce hepatic DNA excision repair in Fischer-344 rats. Covalent binding of the structurally related hepatocarcinogen, 2,6-dinitrotoluene, to hepatic DNA is markedly decreased by prior administration of
D J Doolittle et al.
Cancer research, 43(6), 2836-2842 (1983-06-01)
The nitrotoluenes failed to induce unscheduled DNA synthesis in in vitro cultures of rat hepatocytes. Because intestinal bacteria are known to be involved in the metabolic activation of other nitroaromatic compounds, the genotoxicity of the nitrotoluenes was evaluated using an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service