Skip to Content
Merck
All Photos(1)

Key Documents

JKA7025

Sigma-Aldrich

4arm-PEG20K

average MN 20,000, Vinylsulfone, thiol reactive

Synonym(s):

4arm-PEG-Vinylsulfone, 4arm-PEG-VS, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2O(CH2CH2O)nCH2CH2SO2CHCH2)4
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

4arm-PEG20K-Vinylsulfone, average Mn 20,000

form

solid

mol wt

average Mn 20,000

reaction suitability

reactivity: thiol reactive

polymer architecture

shape: 4-arm
functionality: homofunctional

Looking for similar products? Visit Product Comparison Guide

General description

4arm PEG Vinylsulfone. Hydrogel PEG. VS binds free thiol groups in aqueous buffer between pH 6.5~8.5 at room temperature.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyburz, K.A., et al.
Acta Biomaterialia, 9(5) (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service