Skip to Content
Merck
All Photos(2)

Key Documents

GF75315719

Yttrium

foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%

Synonym(s):

Yttrium, Y 000205

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
MDL number:
UNSPSC Code:
12141617
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 753-157-19

resistivity

57 μΩ-cm, 20°C

size × thickness

25 x 25 mm × 0.005 mm

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Scholz et al.
Dalton transactions (Cambridge, England : 2003), 44(30), 13522-13529 (2015-07-03)
Nanoscopic yttrium acetate fluorides Y(CH(3)COO)(3-z)F(z) and yttrium oxide fluorides YO(3-z)/(2)F(z )were prepared with tunable Y/F molar ratios via the fluorolytic sol-gel route. All samples were characterized by X-ray diffraction, elemental analysis and thermal analysis. In addition, local structures of all
Aki Aoyama et al.
Molecular cancer therapeutics, 13(12), 2978-2990 (2014-10-15)
Tivantinib (ARQ197) was first reported as a highly selective inhibitor of c-MET and is currently being investigated in a phase III clinical trial. However, as recently reported by us and another group, tivantinib showed cytotoxic activity independent of cellular c-MET
Yoshiko Fujita et al.
Environmental science & technology, 49(16), 9460-9468 (2015-07-02)
Increasing rare earth element (REE) supplies by recycling and expanded ore processing will result in generation of new wastewaters. In some cases, disposal to a sewage treatment plant may be favored, but plant performance must be maintained. To assess the
Xue Wang et al.
Journal of nanoscience and nanotechnology, 14(5), 3910-3913 (2014-04-17)
Nd3+ doped alpha-NaYF4 nanocrystals with size of about 15 nm were successfully synthesized through hydrothermal method. Nearly pure near-infrared to near-infrared (NIR-to-NIR) luminescence can be realized. Moreover, the excitation and the emission at 880 and 1060 nm, respectively, are away
[Use of lasers on aluminum-yttrium garnet with neodymium in clinical practice].
O K Skobelkin et al.
Khirurgiia, (8)(8), 155-157 (1990-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service