G7305
Glycoluril
97%
Synonym(s):
Acetylene carbamide, Acetylenediurea, NSC 2765, Tetrahydroimidazo[4,5-d]imidazole-2,5-dione
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C4H6N4O2
CAS Number:
Molecular Weight:
142.12
Beilstein:
128826
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
>300 °C (lit.)
SMILES string
O=C1NC2NC(=O)NC2N1
InChI
1S/C4H6N4O2/c9-3-5-1-2(7-3)8-4(10)6-1/h1-2H,(H2,5,7,9)(H2,6,8,10)
InChI key
VPVSTMAPERLKKM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reactant involved in synthesis of:
- Glycouril hexamers and monofunctionalized cucurbit[6]uril derivatives
- N-chloro compounds from trichloroisocyanuric acid
- Glycouril trimers
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Encapsulated carboxylic acid dimers with compressed hydrogen bonds.
Dariush Ajami et al.
Angewandte Chemie (International ed. in English), 50(2), 528-531 (2010-12-15)
Darren W Johnson et al.
Chemical communications (Cambridge, England), (19)(19), 2228-2229 (2002-10-26)
A series of substituted glycoluril molecules exhibits a substantial twist of the fused five-membered rings and assembles exclusively chiral hydrogen-bonded ribbons in the solid-state.
Dariush Ajami et al.
Topics in current chemistry, 319, 57-78 (2011-11-04)
Synthetic receptors that surround their target molecules - self assembled capsules and deep cavitands - have emerged as the most realistic models of enzymes active sites. They were introduced to study the behaviour of molecules isolated in small spaces and
Longer guests drive the reversible assembly of hyperextended capsules.
Dariush Ajami et al.
Angewandte Chemie (International ed. in English), 46(48), 9283-9286 (2007-11-08)
Neng-Fang She et al.
Journal of the American Chemical Society, 131(33), 11695-11697 (2009-09-03)
A programmed assembly process produces rhombic grid networks from compounds L1-L4 in the crystal by pi-pi stacking interactions that generate a bimolecular grid synthon, which undergoes further NH...N hydrogen-bond-mediated assembly.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service