Skip to Content
Merck
All Photos(2)

Documents

E8506

Sigma-Aldrich

3-(N-Ethyl-3-methylanilino)propanesulfonic acid sodium salt

Synonym(s):

3-(N-Ethyl-m-toluidino)propanesulfonic acid sodium salt, 3-(Ethyl[3-methylphenyl]amino)-1-propanesulfonic acid, N-Ethyl-N-sulfopropyl-m-toluidine sodium salt, Sodium 3-(N-ethyl-3-methylanilino)propanesulfonate, Sodium N-ethyl-N-(3-sulfopropyl)-m-toluidine, TOPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18NNaO3S
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

SMILES string

[Na+].CCN(CCCS([O-])(=O)=O)c1cccc(C)c1

InChI

1S/C12H19NO3S.Na/c1-3-13(8-5-9-17(14,15)16)12-7-4-6-11(2)10-12;/h4,6-7,10H,3,5,8-9H2,1-2H3,(H,14,15,16);/q;+1/p-1

InChI key

NJZLCEFCAHNYIR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis M Kivlighan et al.
Journal of counseling psychology, 63(2), 149-161 (2015-12-23)
We used the Actor Partner Interdependence Model (APIM; Kashy & Kenny, 2000) to examine the dyadic associations of 74 clients and 23 therapists in their evaluations of working alliance, real relationship, session quality, and client improvement over time in ongoing
Dennis M Kivlighan et al.
Journal of counseling psychology, 64(4), 394-409 (2017-04-07)
We examined how congruence and discrepancy in clients' and therapists' ratings of the working alliance (WA) and real relationship (RR) were related to client-rated session quality (SES; Session Evaluation Scale). Ratings for 2517 sessions of 144 clients and 23 therapists
Clara E Hill et al.
Psychotherapy research : journal of the Society for Psychotherapy Research, 24(5), 523-537 (2014-01-07)
Our objectives were to describe the frequency of therapists' dreams about their clients and clients' dreams about their therapists, to determine how therapists and clients who had such dreams differed from those who did not have such dreams, whether therapy
Morgan Anvari et al.
Psychotherapy research : journal of the Society for Psychotherapy Research, 30(7), 900-911 (2019-10-23)
Objective: We wanted to determine whether four therapist skills were differentially associated with emotional expression. Method: We compared paraphrases (restatements and reflections of feelings) and open questions (focused on thoughts or feelings) in relation to antecedent and subsequent client emotional
Hyun Ho Kim et al.
Journal of clinical neurology (Seoul, Korea), 15(4), 502-510 (2019-10-09)
Benign epilepsy with centrotemporal spikes (BECTS) is one of the most common pediatric epilepsies, and it generally has a good prognosis. However, recent research has indicated that the epileptic activity of BECTS can cause cognitive defects such as language, visuospatial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service