Skip to Content
Merck
All Photos(3)

Key Documents

D5765

Sigma-Aldrich

Denatonium benzoate

≥98%

Synonym(s):

N,N-Diethyl-N-[(2,6-dimethylphenyl­carbamoyl)­methyl]­benzyl­ammonium benzoate, Benzyldiethyl(2,6-xylyl­carbamoyl­methyl)­ammonium benzoate, THS 839

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H29N2O · C7H5O2
CAS Number:
Molecular Weight:
446.58
Beilstein:
8179408
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

mp

164-168 °C (lit.)

SMILES string

[O-]C(=O)c1ccccc1.CC[N+](CC)(CC(=O)Nc2c(C)cccc2C)Cc3ccccc3

InChI

1S/C21H28N2O.C7H6O2/c1-5-23(6-2,15-19-13-8-7-9-14-19)16-20(24)22-21-17(3)11-10-12-18(21)4;8-7(9)6-4-2-1-3-5-6/h7-14H,5-6,15-16H2,1-4H3;1-5H,(H,8,9)

InChI key

VWTINHYPRWEBQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Denatonium benzoate (DB) is an ionized compound made up of a negatively charged benzoic acid and quaternary ammonium cation (denatonium). It is used as an alcohol denaturant and flavor in pharmaceuticals. Additionally, used in products like soap, animal repellents, antifreeze.

Application

Denatonium benzoate is used:

  • To prepare anti-gnawing bio-polymer composites using zinc stearate (ZnSt) and capsicum oleoresin (CO) impregnated in silica (SiCO) and polybutylene succinate(PBS).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anne Brockhoff et al.
Journal of agricultural and food chemistry, 55(15), 6236-6243 (2007-06-28)
Sesquiterpene lactones are a major class of natural bitter compounds occurring in vegetables and culinary herbs as well as in aromatic and medicinal plants, where they often represent the main gustatory and pharmacologically active component. Investigations on sesquiterpene lactones have
Effect of capsicum oleoresin and denatonium benzoate in polybutylene succinate on anti-gnawing, thermal and rheological properties
Prasarnsri A, et al.
Materials Today: Proceedings (2022)
W Klein-Schwartz
Veterinary and human toxicology, 33(6), 545-547 (1991-12-01)
The efficacy and toxicity studies on denatonium benzoate are limited and may be subject to varying interpretations when viewed in the context of a potential poisoning situation. Efficacy studies to date in children have shown that in a controlled environment
S R Hansen et al.
Veterinary and human toxicology, 35(3), 234-236 (1993-06-01)
When ingested at 10 ppm by human beings, denatonium benzoate has an extremely bitter, unpleasant taste. The addition of denatonium benzoate to liquid dish detergents and orange juice reduces the amount ingested by children. The toxicity of denatonium benzoate is
Karel Talavera et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 22(5), 1343-1355 (2007-12-12)
Ordinary gustatory experiences, which are usually evoked by taste mixtures, are determined by multiple interactions between different taste stimuli. The most studied model for these gustatory interactions is the suppression of the responses to sweeteners by the prototype bitter compound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service