Skip to Content
Merck
All Photos(1)

Key Documents

CLS6796200

Corning® hotplate, 4 in. x 5 in.

AC/DC input 230 V AC, 50 Hz, 1.1 A (CEE7-7 plug), plate W × L 4 in. × 5 in.

Synonym(s):

stirrers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102406

material

ceramic plate

sterility

non-sterile

packaging

case of 1

manufacturer/tradename

Corning 6796-200

availability

not available in USA, Canada and EU

parameter

25-550 °C temp. range

plate W × L

4 in. × 5 in.

color

none

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

These hot plates have a durable glass-ceramic 4 x 5 inch top and offer superior performance and reliability in a low profile, space-saving design. They are designed to operate on 230V/50Hz with CEE7-7 plug for use in parts of Asia, Africa, Australia and other areas with similar electrical requirements. Easy-to-read numbers simplify measurements, and rugged, extra-large knobs provide greater control for gloved fingers. Hot top indicator light blinks a warning if the top is too hot to touch (>140°F) even when the unit is turned off. Built-in holder accommodates optional support rods. Flatter, whiter top conducts heat more efficiently and evenly. Built-in holder accommodates optional 4 inch stainless steel support rods (No. 400084). Two-year warranty.

Quality

Not CE compliant

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service