Key Documents
CLS4519134
Corning® Probe Insertion Fittings for Direct Overhead Drive 120 mm Center Cap Assembly for 8 L & 15 L Proculture® Spinner Flasks
tubing diam. 1/4 in.
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
sterility
non-sterile
feature
2 tubes
manufacturer/tradename
Corning 4519134
cap diam.
120 mm
tubing diam.
1/4 in.
Other Notes
Cap is sold separately
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries