Skip to Content
Merck
All Photos(2)

Documents

B8703

Sigma-Aldrich

Benzoin methyl ether

96%

Synonym(s):

α-Methoxy-α-phenylacetophenone, 2-Methoxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)COC6H5
CAS Number:
Molecular Weight:
226.27
Beilstein:
2052112
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

crystals

mp

47.5-48.5 °C (lit.)

SMILES string

COC(C(=O)c1ccccc1)c2ccccc2

InChI

1S/C15H14O2/c1-17-15(13-10-6-3-7-11-13)14(16)12-8-4-2-5-9-12/h2-11,15H,1H3

InChI key

BQZJOQXSCSZQPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Photoinitiator.

Other Notes

remainder benzoin

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Droux et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 968, 22-25 (2014-05-13)
We report here the study of the stability under subcritical water conditions of one of the most popular polysaccharide chiral stationary phase (CSP): Chiralcel OD. This CSP was used under high temperature and reversed phase conditions with acetonitrile and 2-propanol
Mengzhe Guo et al.
Analytica chimica acta, 865, 31-38 (2015-03-04)
Matrix-assisted laser desorption ionization (MALDI) time-of-flight mass spectrometry (TOF-MS) can be considered as state of the art in the field of proteins and peptides analysis. In this work, we have designed an ionic liquid derivative strategy to obtain abundant fragment
Benjamin D Brooks et al.
PloS one, 10(3), e0118696-e0118696 (2015-03-31)
Nearly 1.3 million total joint replacement procedures are performed in the United States annually, with numbers projected to rise exponentially in the coming decades. Although finite infection rates for these procedures remain consistently low, device-related infections represent a significant cause
Kira A Ivanova et al.
Protoplasma, 252(6), 1505-1517 (2015-03-07)
Rhizobia are able to establish a beneficial interaction with legumes by forming a new organ, called the symbiotic root nodule, which is a unique ecological niche for rhizobial nitrogen fixation. Rhizobial infection has many similarities with pathogenic infection and induction
Samim Sardar et al.
Scientific reports, 5, 17313-17313 (2015-11-28)
Energy harvesting from solar light employing nanostructured materials offer an economic way to resolve energy and environmental issues. We have developed an efficient light harvesting heterostructure based on poly(diphenylbutadiyne) (PDPB) nanofibers and ZnO nanoparticles (NPs) via a solution phase synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service