ALD00028
4-(Bromomethyl)benzenesulfonyl fluoride
97%
Synonym(s):
4-(Fluorosulfonyl)benzyl bromide, Fluorosulfonylbenzyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H6BrFO2S
CAS Number:
Molecular Weight:
253.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
reaction suitability
reaction type: click chemistry
mp
80-85 °C
SMILES string
O=S(C1=CC=C(CBr)C=C1)(F)=O
InChI
1S/C7H6BrFO2S/c8-5-6-1-3-7(4-2-6)12(9,10)11/h1-4H,5H2
InChI key
WWAPDVQUZYCNCR-UHFFFAOYSA-N
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service