920355
Lithium Tetrafluoroaluminate
98%
Synonym(s):
Aluminum lithium fluoride, Lithium fluoroaluminate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
LiAlF4
CAS Number:
Molecular Weight:
109.92
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
98%
Recommended Products
Quality Level
Assay
98%
InChI
1S/Al.2FH.2F.Li/h;2*1H;;;/q+2;;;;;/p-2
InChI key
GNYGZLVQMJBEMD-UHFFFAOYSA-L
Related Categories
Application
LiAlF4 can be used as a conductive interfical layer for stable cathode cycling. Heterostructured LiAlF4 coatings have shown also promise in improving cycling stability. Lithium tetrafluoroaluminate has been incorporatied into solid electrolyte thin films.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shuoqing Zhao et al.
ACS applied materials & interfaces, 10(39), 33260-33268 (2018-09-07)
Lithium-rich oxides have been regarded as one of the most competitive cathode materials for next-generation lithium-ion batteries due to their high theoretical specific capacity and high discharge voltage. However, they are still far from being commercialized due to low rate
Electrochromism of WO3/LiAlF4/LiIn thin-film overlayers.
Miyauchi K, et al.
Journal of Applied Physics, 53, 1823-1823 (1982)
Jin Xie et al.
ACS nano, 11(7), 7019-7027 (2017-07-01)
Modern lithium ion batteries are often desired to operate at a wide electrochemical window to maximize energy densities. While pushing the limit of cutoff potentials allows batteries to provide greater energy densities with enhanced specific capacities and higher voltage outputs
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service