Skip to Content
Merck
All Photos(1)

Key Documents

915262

Sigma-Aldrich

ZrO2 nanoparticles

5 nm (core), methacrylate functionalized, 50 wt. % in propylene glycol methyl ether acetate (PGMEA), medium surface coverage capping materials

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

Quality Level

form

liquid

concentration

50 wt. % in propylene glycol methyl ether acetate (PGMEA)

particle size

5 nm (core)

functional group

methacrylate

InChI

1S/2O.Zr

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High refractive index nanocrystal dispersions dramatically improve optical and mechanical properties to enable next-generation products. Incorporating ZrO2 significantly increases the refractive index of many commonly used polymers, while maintaining high transparency. This combination of key properties allows researchers to optimize optical efficiency and improve light extraction.

Features and Benefits

  • Zirconia with 5nm core plus surface capping that contains methacrylate functionality designed to crosslink with methacrylate and acrylate systems.
  • Medium surface coverage of capping materials that provides improved dispersibility and high refractive index.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

79.7 °F

Flash Point(C)

26.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service