Skip to Content
Merck
All Photos(1)

Documents

901791

Sigma-Aldrich

Tungsten disulfide suspension

Synonym(s):

2D-Tungsten disulfide, 2D-Tungsten disulfide dispersion with nonionic surfactant, 2D-Tungsten sulfide dispersion, Tungsten sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WS2
UNSPSC Code:
12352302
NACRES:
NA.23

description

50-300 nm
thickness < 3 layers

form

liquid

concentration

1 mg/mL in H2O

Looking for similar products? Visit Product Comparison Guide

General description

Tungsten disulfide suspension (WS2) (1 mg/mL in water) is a two dimensional (2D) transition metal dichalcogenide (TMD) material with a hexagonal configuration. It is a graphene like material that has a honeycomb network. Its properties include high flexibility, and moderate mobility of charge carriers.

Application

Owing to the thickness dependent physical and chemical properties, the transition metal dichalcogenide (TMD) based two dimensional (2D) nanomaterials have drawn significant attention recently. The atomically thin layers of MoS2 and WS2 represent important class of TMD based 2D-material and they find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc.
WS2 suspensions can be used in the fabrication of a variety of devices which include humidity sensors, and lithium ion battery. It can also be used as a catalyst for water decomposition.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic performance of porous laminated tungsten disulfide/copper oxide/single-wall carbon nanotubes hybrids for lithium ions batteries.
Liu Y, et al.
Electrochimica Acta, 148(8), 73-78 (2014)
Tungsten disulfide wrapped on micro fiber for enhanced humidity sensing.
Guan H, et al.
Optical Materials Express, 7(5), 1686-1696 (2017)
Tungsten disulfide (WS 2) based all-fiber-optic humidity sensor.
Luo Y, et al.
Optics Express, 24(8), 8956-8966 (2016)
Tungsten disulfide: a novel hydrogen evolution catalyst for water decomposition.
Sobczynski A, et al.
The Journal of Physical Chemistry, 92(8), 2311-2315 (1988)
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar J L, et al.
The Journal of Physical Chemistry C, 118, 3847-3847 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service