Skip to Content
Merck
All Photos(1)

Key Documents

901138

Sigma-Aldrich

14-Azido-3,6,9,12-tetraoxatetradecan-1-amine

Synonym(s):

2-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethoxy]ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N4O4
CAS Number:
Molecular Weight:
262.31
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: linker

refractive index

n/D 1.45

density

1.10 g/mL

functional group

amine
azide

storage temp.

2-8°C

SMILES string

NCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C10H22N4O4/c11-1-3-15-5-7-17-9-10-18-8-6-16-4-2-13-14-12/h1-11H2

InChI key

ZMBGKXBIVYXREN-UHFFFAOYSA-N

Application

14-Azido-3,6,9,12-tetraoxatetradecan-1-amine is an azide with polyethylene glycol-like characteristics that can be used to prepare fluorescent polymer particles via enzymatic miniemulsion polymerization and copper (I) catalyzed click reaction approach. It is also used as a reactant to synthesize α-aryl-α-diazoamides by aminolysis of N-succinimidyl α-aryl-α-diazoacetates.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A green approach for the synthesis of fluorescent polymer particles by combined use of enzymatic miniemulsion polymerization with clickable surfmer and click reaction
Kohri Michinari, et al.
Transactions of the Materials Research Society of Japan, 39(1), 57-60 (2014)
Joomyung V Jun et al.
Organic letters, 23(8), 3110-3114 (2021-04-06)
α-Aryl-α-diazoamides were synthesized in two steps under mild conditions. This expeditious route employs Pd-catalyzed C-H arylation of N-succinimidyl 2-diazoacetate to obtain N-succinimidyl 2-aryl-2-diazoacetates, followed by aminolysis. The ensuing diazo compounds can esterify carboxyl groups in aqueous solution, and the ester

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service