Skip to Content
Merck
All Photos(1)

Key Documents

900815

Sigma-Aldrich

SLAP 5-TMS TM

≥95%

Synonym(s):

2-((Bis(trimethylsilyl)methyl)thio)ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H25NSSi2
CAS Number:
Molecular Weight:
235.54
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

suspension

density

0.9126

storage temp.

2-8°C

SMILES string

NCCSC([Si](C)(C)C)[Si](C)(C)C

Application

When used with aliphatic or aromatic aldehydes, this silicon amine protocol SLAP reagent enables the photomediated synthesis of thiomorpholines and thiazepanes. For photocatalytic cross-coupling with this SLAP Reagent, Ir[(ppy)2dtbbpy]PF6 (747769) is used with a recommended starting combination of Lewis acids Bi(OTf)3 (633305) and Cu(OTf)2 (283673) prior to subsequent substrate-specific optimization. This product was introduced in collaboration with the Bode Research Group and provides a tin-free alternative to SnAP (tin amine protocol) reagents and is well-suited for scale-up reactions.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.0 °F

Flash Point(C)

-1.11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Ying Hsieh et al.
ACS central science, 3(1), 66-72 (2017-02-06)
Redox neutral photocatalytic transformations often require careful pairing of the substrates and photoredox catalysts in order to achieve a catalytic cycle. This can limit the range of viable transformations, as we recently observed in attempting to extend the scope of
Moritz K Jackl et al.
Organic letters, 19(17), 4696-4699 (2017-08-17)
Photocatalytic coupling of aldehydes and silicon amine protocol (SLAP) reagents enables the simple, scalable synthesis of substituted morpholines, oxazepanes, thiomorpholines, and thiazepanes under continuous flow conditions. Key to the success of this process is the combination of an inexpensive organic
Sheng-Ying Hsieh et al.
Organic letters, 18(9), 2098-2101 (2016-04-22)
Silicon amine protocol (SLAP) reagents for photocatalytic cross-coupling with aldehydes and ketones to form N-unprotected piperazines have been developed. This blue light promoted process tolerates a wide range of heteroaromatic, aromatic, and aliphatic aldehydes and structurally and stereochemically complex SLAP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service