Skip to Content
Merck
All Photos(4)

Key Documents

900445

Sigma-Aldrich

Graphene, monolayer film

1 in x 1 in on copper foil, with PMMA coating, avg. no. of layers, 1

Synonym(s):

PMMA/Graphene/Cu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

product name

Monolayer graphene film, 1 in x 1 in on copper foil, with PMMA coating, avg. no. of layers, 1

Quality Level

description

Film Coverage: >98%
Monolayer coverage: ≥92% (optical)

form

film

quality

Raman analysis-monolayer

feature

avg. no. of layers 1

impurities

<2% (optical)

Looking for similar products? Visit Product Comparison Guide

General description

A single-layer graphene film on a copper substrate.
This product is provided with a PMMA coating on top of the graphene in order to ease the transfer process and to avoid contamination. It′s a high quality product with great homogeneity.

Graphene film
  • Growth method: CVD synthesis
  • Appearance: Transparent film
  • Transparency: > 97%
  • Coverage: > 95%
  • Number of graphene layers: 1
  • Thickness (theoretical): 0.345 nm
  • FET Electron Mobility on Al2O3: 2000 cm2/Vs
  • FET Electron Mobility on SiO2/Si: 4000 cm2/Vs
  • Sheet Resistance on SiO2/Si: 450±40 Ω/sq (1cm x1cm)
  • Grain size: Up to 10 μm

Substrate Cu Foil
  • Thickness: 18 μm
  • Pre-treated for easier bottom layer removal: monolayer graphene on the back side of copper is partially removed, but not completely. Therefore, an additional treatment like RIE is needed before transfer to eliminate the bottom layer totally.

PMMA Coating
  • Spin coated for high homogeneity
  • Molecular weight: 495k
  • Thickness: <100 nm
  • PMMA model: 495K, A2

Application

  • Graphene research
  • Flexible batteries
  • Electronics
  • Aerospace industry
  • MEMS and NEMS
  • Micro actuators
  • Conductive coatings

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and Electrochemical Characterization of Single and Multi-Layer Graphene Anodes for Lithium-Ion Batteries.
Radhakrishnan G,et al.
Journal of the Electrochemical Society, 159(6), A752 - A761 (2012)
Ultrathin rechargeable all-solid-state batteries based on monolayer graphene.
Wei D, et al.
Journal of Material Chemistry A, 1, 3177-3181 (2013)
Xuesong Li et al.
Science (New York, N.Y.), 324(5932), 1312-1314 (2009-05-09)
Graphene has been attracting great interest because of its distinctive band structure and physical properties. Today, graphene is limited to small sizes because it is produced mostly by exfoliating graphite. We grew large-area graphene films of the order of centimeters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service