Skip to Content
Merck
All Photos(2)

Key Documents

900150

Sigma-Aldrich

Cyanomethyl (3,5-Dimethyl-1H-pyrazole)-carbodithioate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3S2
Molecular Weight:
211.31
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

powder

storage condition

protect from light

mp

104-109 °C

SMILES string

CC1=NN(C(SCC#N)=S)C(C)=C1

InChI

1S/C8H9N3S2/c1-6-5-7(2)11(10-6)8(12)13-4-3-9/h5H,4H2,1-2H3

InChI key

OMTFYPHYESIDFH-UHFFFAOYSA-N

Application

A stable, dithiocarbamate chain transfer agent (CTA), also referred to as a RAFT agent, that can control chain growth in free radical polymerization, producing polymers with well-defined molecular weights, and low polydispersities. Cyanomethyl (3,5-Dimethyl-1H-pyrazole)-carbodithioate can be used to control homo-polymerisation and co-polymerisation of acrylic acid, acrylates, acrylamides, and styrenes (2o MAMs), and LAMs such as vinyl acetate and N-vinylpyrrolidinone. It can be used to make MAM-LAM block copolymers. It is soluble in organic media.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense
Patents: WO98/01478, WO99/311444.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dithiocarbamate RAFT agents with broad applicability ? the 3,5-dimethyl-1H-pyrazole-1-carbodithioates.
Gardiner J, et al.
Polym. Chem., 7, 481-492 (2016)
Beth Marbois et al.
The Journal of biological chemistry, 285(36), 27827-27838 (2010-07-02)
Coenzyme Q (ubiquinone or Q) is a crucial mitochondrial lipid required for respiratory electron transport in eukaryotes. 4-Hydroxybenozoate (4HB) is an aromatic ring precursor that forms the benzoquinone ring of Q and is used extensively to examine Q biosynthesis. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service