Skip to Content
Merck
All Photos(3)

Key Documents

856568

Sigma-Aldrich

Polyvinylpyrrolidone

powder, average Mw ~55,000

Synonym(s):

PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mw ~55,000

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PVP has biocompatibility, low toxicity, adhesive characteristics, complexing stability, relatively inert behavior, and is resistant to thermal degradation. It is readily prepared by the polymerization of N-vinyl-2-pyrrolidone or by Reppe synthesis technique.

Application

PVP is one of the constituents used in the synthesis and coating of silver nanocubes. It serves as a reducing agent and colloidal stabilizer in the synthesis of palladium nanobars, triangular,nanoplates,twinned nanorods and bipyramids. It is also used as a capping agent in the synthesis of silver nanospheres,gold nanoframes. General applications of PVP are: in pharmaceutical, food, beverage, cosmetic, toiletry and photographic industries.

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of fivefold twinned nanorods and right
bipyramids of palladium
Xiong Y, et al.
Chemical Physics Letters, 440, 273-278 (2007)
Robinson BV, et al. et al.
PVP: A critical review of the kinetics and toxicology of polyvinlypyrrolidone (Povidone) (1990)
Rapid synthesis of small silver nanocubes by mediating polyol reduction with a trace amount of sodium sulfide or sodium hydrosulfide.
Siekkinen AR, et al.
Chemical Physics Letters, 432(4-6), 491- 496 null
Gold nanocages for cancer imaging and therapy
Au L, et al.
Cancer Nanotechnology, 83-99 (2010)
Synthesis and characterization of fivefold twinned nanorods and right bipyramids of palladium
Xiong Y, et al.
Chemical Physics Letters, 440(4), 273-278 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service